Learn More
Gibco™ MEM α, no nucleosides
MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells.
$84.25 - $758.00
Spécifications
| Classification | Animal Origin-free |
|---|---|
| Concentration | 1 X |
| Form | Liquid |
| Product Type | MEM α (Minimum Essential Medium α) |
| Serum Level | Standard Serum Supplementation |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
12561056
|
Gibco™
12561056 |
500 mL |
chaque for $84.25
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
12561072
|
Gibco™
12561072 |
10 x 500 mL |
chaque for $758.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
12561049
|
Gibco™
12561049 |
1000 mL |
chaque for $158.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This MEM α is modified as follows:
With: Phenol Red, L-glutamine
Without: Ribonucleosides, Deoxyribonucleosides
Using DMEM α
MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR-negative cells. This product is made with Earle's salts. MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Spécifications
| Animal Origin-free | |
| Liquid | |
| Standard Serum Supplementation | |
| Sterile-filtered | |
| Gibco |
| 1 X | |
| MEM α (Minimum Essential Medium α) | |
| Sterile-filtered | |
| cGMP-compliant under the ISO 13485 standard |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.