missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ MEM α, no nucleosides DFS Item

MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells.

Supplier:  Gibco™ 12561056

 View more versions of this product

Catalog No. 12561056


Explore available promotions

Description

Description

MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM α is modified as follows:

With: Phenol Red, L-glutamine

Without: Ribonucleosides, Deoxyribonucleosides

Using DMEM α

MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR-negative cells. This product is made with Earle's salts. MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Primary Rat Astrocytes
1 X
MEM α (Minimum Essential Medium α)
Sterile-filtered
Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
500 mL
HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No HEPES, No Ribonucleosides, No Deoxyribonucleosides
Gibco™
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.