Learn More
Gibco™ PB-MAX™ Karyotyping Medium DFS Item
Fully supplemented medium developed for short-term culture of peripheral blood lymphocytes for cytogenetic studies and in vitro diagnostic procedures
Supplier:  Gibco™ 12557013
 View more versions of this product
Catalog No. 12557013
Only null left
Explore available promotions
Description
Description
  • Ready-to-use format
  • Quality and performance testing
  • Classic formulation
  • Sustainable packaging
Ready-to-use format
1X medium is a frozen, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2° to 8°C for up to 14 days.
Quality and performance testing
Every lot of is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Peripheral blood lymphocytes from a normal adult donor are cultured for 72 hours in Gibco PB-MAX Karyotyping Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot is tested for sterility, pH, osmolality, and mycoplasma.
Classic formulation
Gibco PB-MAX Karyotyping Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), L-glutamine, and phytohemagglutinin (PHA). Gibco PB-MAX Karyotyping Medium is based on formulations referenced in the Association for Cytogenetic Technologists Laboratory Manual for the culture of peripheral blood lymphocytes for cytogenetic analysis.
Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Karyotyping, Mammalian Cell Culture
Specifications
Specifications
Karyotyping Medium
1 X
Liquid
100 mL
L-Glutamine
Red
Cell Viability; Proliferation & Function; Clinical & Translational Research; Mammalian Cell Culture
PB-MAX
Sterile-filtered
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For in vitro diagnostic use.