missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ PB-MAX™ Karyotyping Medium DFS Item

Fully supplemented medium developed for short-term culture of peripheral blood lymphocytes for cytogenetic studies and in vitro diagnostic procedures

$137.00 - $562.00

Specifications

Additive L-Glutamine
Color Red
Content And Storage Storage conditions: -5 to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Dilution 1 X
Product Line PB-MAX™
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12557013 DFS Item
View Documents
Gibco™
12557013
100 mL
Each for $137.00
 
12557021 DFS Item
View Documents
Gibco™
12557021
500 mL
Each for $562.00
 
Description

Description

  • Ready-to-use format
  • Quality and performance testing
  • Classic formulation
  • Sustainable packaging
Ready-to-use format
1X medium is a frozen, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2° to 8°C for up to 14 days.
Quality and performance testing
Every lot of is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Peripheral blood lymphocytes from a normal adult donor are cultured for 72 hours in Gibco PB-MAX Karyotyping Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot is tested for sterility, pH, osmolality, and mycoplasma.
Classic formulation
Gibco PB-MAX Karyotyping Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), L-glutamine, and phytohemagglutinin (PHA). Gibco PB-MAX Karyotyping Medium is based on formulations referenced in the Association for Cytogenetic Technologists Laboratory Manual for the culture of peripheral blood lymphocytes for cytogenetic analysis.

Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Karyotyping, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Compliance

Manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Specifications

L-Glutamine
Storage conditions: -5 to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
PB-MAX™
Human
Sustainable packaging
Standard Serum
Sterile-filtered
Red
1 X
Karyotyping Medium
Cell Viability; Proliferation & Function; Clinical & Translational Research; Mammalian Cell Culture
Lymphocytes
Dry Ice
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For in vitro diagnostic use.