missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ PB-MAX™ Karyotyping Medium DFS Item

Fully supplemented medium developed for short-term culture of peripheral blood lymphocytes for cytogenetic studies and in vitro diagnostic procedures

Supplier:  Gibco™ 12557021

 View more versions of this product

Catalog No. 12557021


Explore available promotions

Description

Description

  • Ready-to-use format
  • Quality and performance testing
  • Classic formulation
  • Sustainable packaging
Ready-to-use format
1X medium is a frozen, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2° to 8°C for up to 14 days.
Quality and performance testing
Every lot of is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Peripheral blood lymphocytes from a normal adult donor are cultured for 72 hours in Gibco PB-MAX Karyotyping Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot is tested for sterility, pH, osmolality, and mycoplasma.
Classic formulation
Gibco PB-MAX Karyotyping Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), L-glutamine, and phytohemagglutinin (PHA). Gibco PB-MAX Karyotyping Medium is based on formulations referenced in the Association for Cytogenetic Technologists Laboratory Manual for the culture of peripheral blood lymphocytes for cytogenetic analysis.

Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Karyotyping, Mammalian Cell Culture

Specifications

Specifications

L-Glutamine
Storage conditions: -5 to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
PB-MAX™
500 mL
Cell Viability; Proliferation & Function; Clinical & Translational Research; Mammalian Cell Culture
Lymphocytes
Dry Ice
Liquid
Red
1 X
Karyotyping Medium
Human
Sustainable packaging
Standard Serum
Sterile-filtered
SDS
Documents

Documents

Promotions

Promotions

For in vitro diagnostic use.