missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Penicillin-Streptomycin (10,000 U/mL)
For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
Supplier: Gibco™ 15140122
Description
Penicillin was originally purified from fungi Penicillium that acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.
- Contains 10,000 units/mL of penicillin
- 10,000μg/mL of streptomycin in a 10 mM citrate buffer (for pH stability)
Specifications
Storage conditions: -5°C to -20°C Shipping conditions: Dry ice Shelf life: 12 months from date of manufacture |
|
Liquid | |
Sterile-filtered | |
Prevention of Cell Culture Contamination | |
Dry Ice |
100 X | |
Antibiotic | |
Sterile-filtered | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.