missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Penicillin-Streptomycin (10,000 U/mL) DFS Item

For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria

$25.82 - $772.00

Specifications

Concentration 100 X
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
15140148 DFS Item
View Documents
Gibco™
15140148
20 mL
Each for $25.82
 
15140122 DFS Item
View Documents
Gibco™
15140122
100 mL
Each for $45.14
 
15140163 DFS Item
View Documents
Gibco™
15140163
20 x 100 mL
Each for $772.00
 
Description

Description

Penicillin was originally purified from fungi Penicillium that acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.

  • Contains 10,000 units/mL of penicillin
  • 10,000μg/mL of streptomycin in a 10 mM citrate buffer (for pH stability)
Specifications

Specifications

100 X
Antibiotic
Sterile-filtered
Liquid
Sterile-filtered
Prevention of Cell Culture Contamination
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.