missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Penicillin-Streptomycin (10,000 U/mL) DFS Item

For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria

Supplier:  Gibco™ 15140163

 View more versions of this product

Catalog No. 15140163


Explore available promotions

Description

Description

Penicillin was originally purified from fungi Penicillium that acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.

  • Contains 10,000 units/mL of penicillin
  • 10,000μg/mL of streptomycin in a 10 mM citrate buffer (for pH stability)
Specifications

Specifications

Storage conditions: -5°C to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Liquid
Sterile-filtered
Prevention of Cell Culture Contamination
20 x 100 mL
100 X
Antibiotic
Sterile-filtered
Gibco™
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.