Gibco™ Distilled Water DFS Item
High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
Supplier:  Gibco™ 15230001
 View more versions of this product
Catalog No. 15230001
Explore available promotions
Description
Description
  • High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
  • Water is prepared by distillation and filtration (0.1 μm), with no added substances
  • Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly
  • Can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Store at 15°C to 30°C
Specifications
Specifications
Cell Culture
Sterile
6 x 1 L
Room Temperature
Bottle
Not DEPC-Treated
Membrane-Filtered
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.