missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Distilled Water
High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
$16.39 - $354.00
Specifications
Grade | Cell Culture |
---|---|
Packaging | Bottle |
Sterility | Sterile |
Treatment(s) | Not DEPC-Treated |
Purification Method | Membrane-Filtered |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
15230170
|
Gibco™
15230170 |
100 mL |
Each for $16.39
|
|
|||||
15230162
|
Gibco™
15230162 |
500 mL |
Each for $41.31
|
|
|||||
15230147
|
Gibco™
15230147 |
1000 mL |
Each for $76.00
|
|
|||||
15230196
|
Gibco™
15230196 |
20 x 100 mL |
Each for $267.00
|
|
|||||
15230204
|
Gibco™
15230204 |
10 x 500 mL |
Each for $354.00
|
|
|||||
15230001
|
Gibco™
15230001 |
6 x 1 L |
Each for $270.00
|
|
|||||
Description
- High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
- Water is prepared by distillation and filtration (0.1 μm), with no added substances
- Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly
- Can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
- Store at 15°C to 30°C
Order Info
Shipping Condition: Room Temperature
Specifications
Cell Culture | |
Sterile | |
Membrane-Filtered |
Bottle | |
Not DEPC-Treated | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.