missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Distilled Water DFS Item

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents

Supplier:  Gibco™ 15230147

 View more versions of this product

Catalog No. 15230147

Explore available promotions

Description

Description

  • High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
  • Water is prepared by distillation and filtration (0.1 μm), with no added substances
  • Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly
  • Can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Store at 15°C to 30°C
Specifications

Specifications

7
Bottle
Not DEPC-Treated
Membrane-Filtered
Cell Culture
Sterile
1000 mL
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.