missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Distilled Water
High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
Supplier: Gibco™ 15230196
Description
- High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
- Water is prepared by distillation and filtration (0.1 μm), with no added substances
- Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly
- Can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
- Store at 15°C to 30°C
Specifications
Cell Culture | |
Sterile | |
20 x 100 mL | |
Room Temperature |
Bottle | |
Not DEPC-Treated | |
Membrane-Filtered |
For Research Use Only. Not for use in diagnostic procedures.