Gilson™ PIPETMAN™ Classic Pipet
The PIPETMAN Classic line, the world standard in pipetting, offers a wide range of continuously adjustable air-displacement pipettes suited for aqueous solutions of moderate viscosity and density.
Supplier:  Gilson™ F144802
 View more versions of this product
Catalog No. F144802G
Explore available promotions
Description
Description
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are autoclavable to avoid cross-contamination
  • Each pipet bears engraved identification number for traceability
Order Info
Trade in program available. Please contact your Fisher Scientific Sales representative for more details.
Specifications
Specifications
±0.025, ±2.5μL
No
Manual
Ergonomic
P10
Pipetman Classic
Non-sterile
1 to 10 μL
14.37 in.
12.01 in.
4.72 in.
Red
D10, DL10, DF10ST, DFL10ST
Red
Reusable
Polypropylene
1
No
Pipette
PCR, DNA Sequencing, Small Volume Dispensing of Aqueous Fluids of Moderate Viscosity and Density (Molecular Biology, Immunology)
36.5 cm
30.5 cm
12 cm
Certified Quality