Gilson™ PIPETMAN™ L Multichannel Pipet
The PIPETMAN™ L Multichannel Pipet is a lightweight multichannel pipet with lockable volume that has been designed to fit naturally in your hand.
Supplier:  Gilson™ FA10012
 View more versions of this product
Catalog No. FA10012G
Explore available promotions
Description
Description
  • Gilson–patented volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • Redesigned handle fits naturally in your hand, no matter the size.
  • Unique internal design applies even, equal pressure from either side of the pipette, rather than on the center of the channels like other multichannels do.
  • 2D identification code tracks product reference number, date of manufacture, serial number and maximum volume of pipet.
  • Available in 8–and 12–channel covering a volume range from 0.5 to 300μL.
Specifications
Specifications
No
Yellow
Reusable
Polypropylene
12
No
Pipette
Ergonomic
6.02 in.
12.87 in.
1.5 in.
0.9 lb.
Certified Quality
Autoclavable
Manual
Ergonomic
P12x200L
Pipetman™ L
Non-sterile
20 to 200 μL
1 Year
15.3 cm
32.7 cm
3.8 cm
Yellow
SDS
missing translation for 'documents'
missing translation for 'documents'
Accessories