Gilson™ PIPETMAN™ L Multichannel Pipet
The PIPETMAN™ L Multichannel Pipet is a lightweight multichannel pipet with lockable volume that has been designed to fit naturally in your hand.
Supplier:  Gilson™ FA10013
 View more versions of this product
Catalog No. FA10013G
Explore available promotions
Description
Description
  • Gilson–patented volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • Redesigned handle fits naturally in your hand, no matter the size.
  • Unique internal design applies even, equal pressure from either side of the pipette, rather than on the center of the channels like other multichannels do.
  • 2D identification code tracks product reference number, date of manufacture, serial number and maximum volume of pipet.
  • Available in 8–and 12–channel covering a volume range from 0.5 to 300μL.
Specifications
Specifications
No
Red
Reusable
Polypropylene
8
No
Pipette
Ergonomic
5.98 in.
12.83 in.
1.54 in.
0.81 lb.
Certified Quality
Autoclavable
Manual
Ergonomic
P8x10L
Pipetman™ L
Non-sterile
0.5 to 10 μL
1 Year
15.2 cm
32.6 cm
3.9 cm
Red
SDS
Documents
Documents
Accessories