missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Glass Bijou Bottles with Cap

Glass Bijou bottles with rubber lined aluminum threaded screw caps for storing liquid solutions.

$6.35 - $9.74

Specifications

Color Clear
For Use With (Application) Solution Storage
Includes Glass Bottle, Aluminum Cap
Material Glass
Quantity 1
View More Specs

Includes: Glass bottle, aluminum cap

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S39311
View Documents
Eisco™
CH0188A
7 mL
Each for $6.35
 
S39312
View Documents
Eisco™
CH0188B
14 mL
Each for $9.74
 
Description

Description

  • Includes glass bottle and aluminum cap
  • Cap is threaded and lined with rubber for a tight leak proof seal

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Clear
Glass Bottle, Aluminum Cap
1
Yes
Reusable
Narrow
Bijou Bottle
Rubber
Solution Storage
Glass
Non-sterile
Autoclavable
No
Cylindrical
Unlabeled
Screw Cap
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'