missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Globe Scientific 4 oz. Specimen Containers

With EZ-Turn quarter turning screw cap designed to assist in prevention of carpal tunnel syndrome

Supplier:  Globe Scientific 5913

 View more versions of this product

Catalog No. 21-200-292


Explore available promotions

Description

Description

  • Polypropylene containers with polyethylene caps
  • Used for collecting urine sample
  • Graduated to 100mL with frosted writing area
  • Comes with trilingual patient ID label
  • Sterile

Specifications

Specifications

Yes
4 oz.
Green
Sterile
Screw Cap
Videos
SDS
Documents

Documents

Promotions

Promotions