missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Globe Scientific 4 oz. Specimen Containers

With EZ-Turn quarter turning screw cap designed to assist in prevention of carpal tunnel syndrome

$86.09 - $309.78

Specifications

Graduated Yes
Closure Type Screw Cap
View More Specs

Products 3
Catalog Number Mfr. No. Color Sterility Price Quantity  
Catalog Number Mfr. No. Color Sterility Price Quantity  
14-377-68 Edge
View Documents
Globe Scientific
5915
Red Non-sterile
Case of 500 Each for $309.78
Edge
 
22-170-143
View Documents
Globe Scientific
5912
Red Sterile
Case of 100 Each for $86.09
 
21-200-292
View Documents
Globe Scientific
5913
Green Sterile
Case of 100 Each for $102.25
 
Description

Description

 

  • Polypropylene containers with polyethylene caps
  • Used for collecting urine sample
  • Graduated to 100mL with frosted writing area
  • Comes with trilingual patient ID label
  • Some units are sterile

 

Specifications

Specifications

Yes
Screw Cap
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'