Globe Scientific Accessories For Sedi-Rate™ ESR Systems
The unique rack (# 3452) features support clamps that hold pipets in a perfectly verticle position. When used in conjunction with the acrylic leveling platform (# 3459), users are assured of the most accurate results. The durable acrylic leveling platform has a leveling bubble and four adjustable feet. It is used in conjunction with the ESR Rack with Support Clamps to ensure a completely leveling testing surface.
Supplier: Globe Scientific 3452
Description
- Ensure ESR Testing Pipettes are perfectly vertical for the most accurate results
Specifications
Rack with Pipette Clamps | |
Waived | |
Sedi-Rate ESR Systems | |
10 |
No | |
ESR Rack | |
Pipette support clamps | |
Each |