Globe Scientific Accessories For Sedi-Rate™ ESR Systems
The unique rack (# 3452) features support clamps that hold pipets in a perfectly verticle position. When used in conjunction with the acrylic leveling platform (# 3459), users are assured of the most accurate results. The durable acrylic leveling platform has a leveling bubble and four adjustable feet. It is used in conjunction with the ESR Rack with Support Clamps to ensure a completely leveling testing surface.
$80.91 - $299.62
Catalog Number | Mfr. No. | Description | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Type | Price | Quantity | |||||
21-200-208
|
Globe Scientific
3452 |
Rack with Pipette Clamps | ESR Rack |
Each for $211.82
|
|
|||||
21-200-209
|
Globe Scientific
3459 |
Acrylic Leveling Support | ESR Leveling Support |
Each for $299.62
|
|
|||||
21-200-210
|
Globe Scientific
3472DUP |
10 Place polystyrene rack | ESR Rack |
Each for $80.91
|
|
|||||
Description
- Ensure ESR Testing Pipettes are perfectly vertical for the most accurate results
Specifications
Waived | |
Each |