Globe Scientific Sterile Rigid 10uL & 1uL Combo Inoculation Loops
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Supplier: Globe Scientific 3850F
Description
- Rigid, medical grade of polystyrene (PS) construction
- 10μL and 1μL
- Sterilized by gamma radiation
- White
- Available individually wrapped or in peel packs
Specifications
Single-use | |
Polystyrene | |
Natural | |
Needle and calibration certificate |
Sterile | |
1 μL | |
Combo Inoculation Loop | |
Individually wrapped |