Globe Scientific Test Tubes
Multi-use test tubes to fit most laboratory applications
Supplier:  Globe Scientific 119040
 View more versions of this product
Catalog No. 21-200-268
Explore available promotions
Description
Description
  • Non-sterile
  • Molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents
  • Polystyrene tubes can withstand 3000rcf
  • Polypropylene tubes can withstand 5000rcf and are autoclavable and freezable
Specifications
Specifications
8 mL
Polypropylene
Yes
No
Test Tube
75 mm
2500/Cs.
Autoclavable
16 mm