Globe Scientific Test Tubes
Multi-use test tubes to fit most laboratory applications
$141.70 - $641.36
Catalog Number | Mfr. No. | Material | Length (Metric) | Diameter (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Length (Metric) | Diameter (Metric) | Price | Quantity | |||
22-170-153
|
Globe Scientific
110445 |
Polypropylene | 100 mm | 13 mm |
Case of 1000 Each for $160.79
|
|
|||
22-170-157
|
Globe Scientific
110415 |
Polystyrene | 100 mm | 13 mm |
Case of 1000 Each for $150.33
|
|
|||
21-200-265
|
Globe Scientific
119010A |
Polystyrene | 75 mm | 16 mm |
Case of 1000 Each for $302.42
|
|
|||
21-200-266
|
Globe Scientific
GLB119010 |
Polystyrene | 75 mm | 16 mm |
Case of 2500 Each for $641.36
|
|
|||
21-200-268
|
Globe Scientific
119040 |
Polypropylene | 75 mm | 16 mm |
Case of 2500 Each for $548.30
|
|
|||
36-101-6879
|
N/A | Polypropylene | 75 mm | 16 mm |
Case of 1000 Each for $275.03
|
|
|||
21-200-269
|
Globe Scientific
117011 |
Polystyrene | 55 mm | 12 mm |
Pack of 1000 Each for $141.70
|
|
|||
21-200-270
|
Globe Scientific
118011 |
Polystyrene | 70 mm | 11 mm |
Pack of 1000 Each for $165.09
|
|
|||
Description
- Non-sterile
- Molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents
- Polystyrene tubes can withstand 3000rcf
- Polypropylene tubes can withstand 5000rcf and are autoclavable and freezable
Specifications
Test Tube |