missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gluconic acid, sodium salt, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 181390100

 View more versions of this product

Catalog No. AC181390100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gluconic acid, sodium salt
206.0°C to 209.0°C
6.5 to 7.5 (10% soln.)
0.98
C6H11NaO7
MFCD00064210
03, I, 188
+ 12.10 (23.00°C c=1.75,H2O)
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]
218.137
CHEBI:84997
0.98
527-07-1
White to Beige
Authentic
Plastic drum
HOCH2[CH(OH)]4CO2Na
10 kg
15, 4492
+ 12.10
UPMFZISCCZSDND-JJKGCWMISA-M
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
23672301
218.14
Crystalline Powder

Chemical Identifiers

527-07-1
218.137
UPMFZISCCZSDND-JJKGCWMISA-M
23672301
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C6H11NaO7
MFCD00064210
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
CHEBI:84997
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only