missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gluconic acid, sodium salt, 98%, Thermo Scientific Chemicals
$57.60 - $764.15
Chemical Identifiers
CAS | 527-07-1 |
---|---|
Molecular Formula | C6H11NaO7 |
Molecular Weight (g/mol) | 218.137 |
MDL Number | MFCD00064210 |
InChI Key | UPMFZISCCZSDND-JJKGCWMISA-M |
Synonym | sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t |
PubChem CID | 23672301 |
ChEBI | CHEBI:84997 |
IUPAC Name | sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
SMILES | C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC181390050
|
Thermo Scientific Chemicals
181390050 |
5 g | Glass Bottle |
Each for $57.60
|
|
||||
AC181390010
|
Thermo Scientific Chemicals
181390010 |
1 kg | Plastic Bottle |
Each for $98.40
|
|
||||
AC181390025
|
Thermo Scientific Chemicals
181390025 |
2.5 kg | Plastic Bottle |
Each for $225.46
|
|
||||
AC181390100
|
Thermo Scientific Chemicals
181390100 |
10 kg | Plastic drum |
Each for $764.15
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
527-07-1 | |
218.137 | |
UPMFZISCCZSDND-JJKGCWMISA-M | |
23672301 | |
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
C6H11NaO7 | |
MFCD00064210 | |
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t | |
CHEBI:84997 | |
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+] |
Specifications
527-07-1 | |
White to Beige | |
Authentic | |
Glass Bottle | |
HOCH2[CH(OH)]4CO2Na | |
5 g | |
15, 4492 | |
+ 12.10 | |
UPMFZISCCZSDND-JJKGCWMISA-M | |
sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate | |
23672301 | |
218.14 | |
Crystalline Powder |
206.0°C to 209.0°C | |
6.5 to 7.5 (10% soln.) | |
0.98 | |
C6H11NaO7 | |
MFCD00064210 | |
03, I, 188 | |
+ 12.10 (23.00°C c=1.75,H2O) | |
sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t | |
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+] | |
218.137 | |
CHEBI:84997 | |
0.98 | |
Gluconic acid, sodium salt |
RUO – Research Use Only