missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaric acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1459514

 View more versions of this product

Catalog No. AAA1459514


Explore available promotions

Description

Description

Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

Solubility
Soluble in water, alcohol, benzene and chloroform. Slightly soluble in petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications

Specifications

Glutaric acid
95°C to 99°C
200°C (20 mmHg)
MFCD00004410
1209725
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
JFCQEDHGNNZCLN-UHFFFAOYSA-N
pentanedioic acid
743
132.12
110-94-1
1.429
C5H8O4
25 g
14,4473
Soluble in water,alcohol,benzene and chloroform. Slightly soluble in petroleum ether.
OC(=O)CCCC(O)=O
132.12
CHEBI:17859
99%

Chemical Identifiers

110-94-1
132.12
JFCQEDHGNNZCLN-UHFFFAOYSA-N
743
pentanedioic acid
C5H8O4
MFCD00004410
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
CHEBI:17859
OC(=O)CCCC(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

EINECSNumber : 203-817-2

RTECSNumber : MA3740000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only