Learn More
Glutaric acid, 99%, Thermo Scientific Chemicals
$48.77 - $389.37
Chemical Identifiers
CAS | 110-94-1 |
---|---|
Molecular Formula | C5H8O4 |
Molecular Weight (g/mol) | 132.12 |
MDL Number | MFCD00004410 |
InChI Key | JFCQEDHGNNZCLN-UHFFFAOYSA-N |
Synonym | glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di |
PubChem CID | 743 |
ChEBI | CHEBI:17859 |
IUPAC Name | pentanedioic acid |
SMILES | OC(=O)CCCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1459514
|
Thermo Scientific Chemicals
A1459514 |
25 g |
Each for $48.77
|
|
|||||
AAA1459522
|
Thermo Scientific Chemicals
A1459522 |
100 g |
Each for $108.58
|
|
|||||
AAA1459536
|
Thermo Scientific Chemicals
A1459536 |
500 g |
Each for $389.37
|
|
|||||
Description
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.
Solubility
Soluble in water, alcohol, benzene and chloroform. Slightly soluble in petroleum ether.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Chemical Identifiers
110-94-1 | |
132.12 | |
JFCQEDHGNNZCLN-UHFFFAOYSA-N | |
743 | |
pentanedioic acid |
C5H8O4 | |
MFCD00004410 | |
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di | |
CHEBI:17859 | |
OC(=O)CCCC(O)=O |
Specifications
110-94-1 | |
1.429 | |
C5H8O4 | |
25 g | |
14,4473 | |
Soluble in water,alcohol,benzene and chloroform. Slightly soluble in petroleum ether. | |
OC(=O)CCCC(O)=O | |
132.12 | |
CHEBI:17859 | |
99% |
95°C to 99°C | |
200°C (20 mmHg) | |
MFCD00004410 | |
1209725 | |
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di | |
JFCQEDHGNNZCLN-UHFFFAOYSA-N | |
pentanedioic acid | |
743 | |
132.12 | |
Glutaric acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
EINECSNumber : 203-817-2
RTECSNumber : MA3740000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only