missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutaric dialdehyde, 25 wt.% solution in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 119980025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glutaric dialdehyde | |
-5.0°C | |
3.2 to 4.2 | |
Glass bottle | |
1.373 | |
MFCD00007025 | |
01, 776 | |
1.06 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.12 | |
Solution |
111-30-8,7732-18-5 | |
1.0600g/mL | |
101°C | |
C5H8O2 | |
OHC(CH2)3CHO | |
2.5 L | |
01,411 | |
15, 4508 | |
Solubility in water: soluble | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
16.4mmHg at 20°C |
RUO – Research Use Only