missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaric dialdehyde, 25 wt.% solution in water, Thermo Scientific Chemicals

$92.06 - $1019.31

Specifications

Chemical Name or Material Glutaric dialdehyde
CAS 111-30-8,7732-18-5
Density 1.0600g/mL
pH 3.2 to 4.2
Molecular Formula C5H8O2
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119980250
View Documents Promotion Details
Thermo Scientific Chemicals
119980250
25 mL Glass bottle
Each for $92.06
 
AC119980010
View Documents
Thermo Scientific Chemicals
119980010
1 L Glass Bottle
Each for $204.71
 
AC119980025
View Documents
Thermo Scientific Chemicals
119980025
2.5 L Glass bottle
Each for $377.31
 
AC119980100
View Documents
Thermo Scientific Chemicals
119980100
10 L Plastic drum
Each for $1,019.31
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glutaric dialdehyde
1.0600g/mL
C5H8O2
MFCD00007025
01,411
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
O=CCCCC=O
100.12
CHEBI:64276
16.4mmHg at 20°C
111-30-8,7732-18-5
3.2 to 4.2
OHC(CH2)3CHO
01, 776
15, 4508
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
pentanedial
3485
100.12
Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only