missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaric dialdehyde, 25 wt.% solution in water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 119980250

 View more versions of this product

Catalog No. AC119980250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glutaric dialdehyde
-5.0°C
3.2 to 4.2
Glass bottle
1.373
MFCD00007025
01, 776
1.06
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
pentanedial
3485
100.12
Solution
111-30-8,7732-18-5
1.0600g/mL
101°C
C5H8O2
OHC(CH2)3CHO
25 mL
01,411
15, 4508
Solubility in water: soluble
O=CCCCC=O
100.12
CHEBI:64276
16.4mmHg at 20°C
Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only