missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaric dialdehyde, 50 wt.% solution in water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 410960025

 View more versions of this product

Catalog No. AC410960025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glutaric dialdehyde
111-30-8,7732-18-5
1.1300g/mL
101.5°C (740.0 mmHg)
C5H8O2
MFCD00007025
01, 776
1.13
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
pentanedial
3485
CHEBI:64276
Solution
50 wt% Solution in Water
-33.0°C
3.2 to 4.2
Glass bottle
OHC(CH2)3CHO
2.5 kg
01,411
15, 4508
Solubility in water: soluble
O=CCCCC=O
100.12
20 mPa.s (50°C)
100.12
Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only