missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutaric dialdehyde, 50 wt.% solution in water, Thermo Scientific Chemicals
$190.65 - $992.00
Specifications
Chemical Name or Material | Glutaric dialdehyde |
---|---|
Name Note | 50 wt% Solution in Water |
CAS | 111-30-8,7732-18-5 |
Melting Point | -33.0°C |
Density | 1.1300g/mL |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC410960010
|
Thermo Scientific Chemicals
410960010 |
1 kg | Glass bottle |
Each for $190.65
|
|
||||
AC410960025
|
Thermo Scientific Chemicals
410960025 |
2.5 kg | Glass bottle |
Each for $240.19
|
|
||||
AC410960100
|
Thermo Scientific Chemicals
410960100 |
10 kg | Plastic drum |
Each for $992.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glutaric dialdehyde | |
111-30-8,7732-18-5 | |
1.1300g/mL | |
101.5°C (740.0 mmHg) | |
OHC(CH2)3CHO | |
01, 776 | |
15, 4508 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
20 mPa.s (50°C) | |
Solution |
50 wt% Solution in Water | |
-33.0°C | |
3.2 to 4.2 | |
C5H8O2 | |
MFCD00007025 | |
01,411 | |
1.13 | |
Solubility in water: soluble | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
100.12 |
RUO – Research Use Only