Learn More
Glutaryl dichloride, 97%
CAS: 2873-74-7 | C5H6Cl2O2 | 169.01 g/mol
$120.34 - $120.34
Identifiants chimiques
| CAS | 2873-74-7 |
|---|---|
| Molecular Formula | C5H6Cl2O2 |
| Molecular Weight (g/mol) | 169.01 |
| MDL Number | MFCD00000755 |
| InChI Key | YVOFTMXWTWHRBH-UHFFFAOYSA-N |
| Synonym | glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride |
| PubChem CID | 17887 |
| IUPAC Name | pentanedioyl dichloride |
| SMILES | C(CC(=O)Cl)CC(=O)Cl |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC119990250
|
Thermo Scientific Chemicals
119990250 |
25 mL | Glass bottle |
chaque for $120.34
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 2873-74-7 | |
| 169.01 | |
| YVOFTMXWTWHRBH-UHFFFAOYSA-N | |
| 17887 | |
| C(CC(=O)Cl)CC(=O)Cl |
| C5H6Cl2O2 | |
| MFCD00000755 | |
| glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride | |
| pentanedioyl dichloride |
Spécifications
| 2873-74-7 | |
| 1.3240g/mL | |
| 106°C | |
| 97% | |
| C5H6Cl2O2 | |
| ClCO(CH2)3COCl | |
| 25 mL | |
| 1.324 | |
| Solubility in water: reacts with water. | |
| C(CC(=O)Cl)CC(=O)Cl | |
| 169.01 | |
| 169.01 | |
| Liquid |
| Brown-Red or Colorless to Yellow | |
| 216.0°C to 218.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4720 to 1.4760 | |
| MFCD00000755 | |
| 02, 634 | |
| glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride | |
| YVOFTMXWTWHRBH-UHFFFAOYSA-N | |
| pentanedioyl dichloride | |
| 17887 | |
| 97% | |
| Glutaryl dichloride, 97% |
Sécurité et manipulation
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-711-1
missing translation for 'rtecsNumber' : SA2583000
missing translation for 'tsca' : TSCA
RUO – Research Use Only