missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerine trioleate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 368121000

 View more versions of this product

Catalog No. AC368121000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glycerine trioleate
Colorless to Yellow
235.0°C to 240.0°C (18.0 mmHg)
98.5% min. (GC)
1.4680 to 1.4700
MFCD00137563
02,IV,1664
15,9904
Solubility in water: insoluble in water. Other solubilities: soluble in chloroform, ether, ccl4, slightly soluble in alcohol
CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC
885.45
885.45
Liquid
122-32-7
0.9130g/mL
Authentic
C57H104O6
(C17H33COOCH2)2CHOCOC17H33
100 mL
0.913
triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate
PHYFQTYBJUILEZ-BSCDBXJPSA-N
[2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
45253964
99%

Chemical Identifiers

122-32-7
885.45
PHYFQTYBJUILEZ-BSCDBXJPSA-N
45253964
CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC
C57H104O6
MFCD00137563
triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate
[2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-534-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only