missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerine trioleate, 98%
CAS: 122-32-7 | C57H104O6 | 885.45 g/mol
$75.27 - $4001.76
Chemical Identifiers
| CAS | 122-32-7 |
|---|---|
| Molecular Formula | C57H104O6 |
| Molecular Weight (g/mol) | 885.45 |
| MDL Number | MFCD00137563 |
| InChI Key | PHYFQTYBJUILEZ-BSCDBXJPSA-N |
| Synonym | triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate |
| PubChem CID | 45253964 |
| IUPAC Name | [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate |
| SMILES | CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC368120010
|
Thermo Scientific Chemicals
368120010 |
1 mL |
Each for $75.27
|
|
|||||
|
AC368120050
|
Thermo Scientific Chemicals
368120050 |
5 mL |
Each for $187.55
|
|
|||||
|
AC368120250
|
Thermo Scientific Chemicals
368120250 |
25 mL |
Each for $861.98
|
|
|||||
|
AC368121000
|
Thermo Scientific Chemicals
368121000 |
100 mL |
Each for $4,001.76
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 122-32-7 | |
| 885.45 | |
| PHYFQTYBJUILEZ-BSCDBXJPSA-N | |
| 45253964 | |
| CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC |
| C57H104O6 | |
| MFCD00137563 | |
| triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate | |
| [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate |
Specifications
| 122-32-7 | |
| 0.9130g/mL | |
| Authentic | |
| Glass bottle | |
| 1.4680 to 1.4700 | |
| MFCD00137563 | |
| 02,IV,1664 | |
| 15,9904 | |
| Solubility in water: insoluble in water. Other solubilities: soluble in chloroform, ether, ccl4, slightly soluble in alcohol | |
| CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC | |
| 885.45 | |
| 885.45 | |
| Liquid |
| Colorless to Yellow | |
| 235.0°C to 240.0°C (18.0 mmHg) | |
| 98.5% min. (GC) | |
| C57H104O6 | |
| (C17H33COOCH2)2CHOCOC17H33 | |
| 1 mL | |
| 0.913 | |
| triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate | |
| PHYFQTYBJUILEZ-BSCDBXJPSA-N | |
| [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate | |
| 45253964 | |
| 99% | |
| Glycerine trioleate |
Safety and Handling
EINECSNumber : 204-534-7
RUO – Research Use Only