missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158922500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glycerol | |
56-81-5 | |
18.0°C | |
290.0°C | |
Authentic | |
Glass bottle | |
1.4716 | |
MFCD00004722 | |
01, 502 | |
15, 4520 | |
Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform | |
OCC(O)CO | |
92.09 | |
1069 mPa.s (20°C) | |
92.09 | |
99+% | |
Viscous Liquid |
Extra Pure | |
>290°C | |
1.2610g/mL | |
160°C | |
99% min. (GC) | |
C3H8O3 | |
HOCH2CH(OH)CH2OH | |
250 mL | |
1.261 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
CHEBI:17754 | |
0.003mbar at 50°C | |
Extra Pure |
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Safety and Handling
EINECSNumber : 200-289-5
RUO – Research Use Only