missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycerol, 99+%, extra pure, Thermo Scientific Chemicals
$42.98 - $820.48
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.09 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | OCC(O)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC158922500
|
Thermo Scientific Chemicals
158922500 |
250 mL | Glass bottle |
Each for $42.98
|
|
||||
AC158920025
|
Thermo Scientific Chemicals
158920025 |
2.5 L | Glass Bottle |
Each for $202.40
Case of 4 Each for $0.00
|
|
||||
AC158920100
|
Thermo Scientific Chemicals
158920100 |
10 L | Plastic drum |
Each for $820.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Specifications
56-81-5 | |
18.0°C | |
290.0°C | |
Authentic | |
Glass bottle | |
1.4716 | |
MFCD00004722 | |
01, 502 | |
15, 4520 | |
Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform | |
OCC(O)CO | |
92.09 | |
1069 mPa.s (20°C) | |
92.09 | |
99+% | |
Viscous Liquid |
>290°C | |
1.2610g/mL | |
160°C | |
99% min. (GC) | |
C3H8O3 | |
HOCH2CH(OH)CH2OH | |
250 mL | |
1.261 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
CHEBI:17754 | |
0.003mbar at 50°C | |
Extra Pure | |
Glycerol, Extra Pure |
Safety and Handling
EINECSNumber : 200-289-5
RUO – Research Use Only