missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycidol, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 120055000

 View more versions of this product

Catalog No. AC120055000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glycidol
-54.0°C
1.1100g/mL
71°C
Glass bottle
1.4300 to 1.4340
500 mL
1.11
glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol
CTKINSOISVBQLD-UHFFFAOYNA-N
oxiran-2-ylmethanol
11164
CHEBI:30966
96%
556-52-5
Beige-Yellow to White
160.0°C to 161.0°C
Authentic
C3H6O2
MFCD00005147,MFCD00074873
17,104
15,4525
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
OCC1CO1
74.08
4.0 mPa.s (20°C)
74.08
Powder, Crystals or Chunks

Chemical Identifiers

556-52-5
74.08
CTKINSOISVBQLD-UHFFFAOYNA-N
11164
oxiran-2-ylmethanol
C3H6O2
MFCD00005147,MFCD00074873
glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol
CHEBI:30966
OCC1CO1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
May cause cancer.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:

GHS Signal Word: Danger

EINECSNumber : 209-128-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only