Learn More
Glycidol, 96%, Thermo Scientific Chemicals
$105.24 - $379.46
Chemical Identifiers
CAS | 556-52-5 |
---|---|
Molecular Formula | C3H6O2 |
Molecular Weight (g/mol) | 74.08 |
MDL Number | MFCD00005147,MFCD00074873 |
InChI Key | CTKINSOISVBQLD-UHFFFAOYNA-N |
Synonym | glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol |
PubChem CID | 11164 |
ChEBI | CHEBI:30966 |
IUPAC Name | oxiran-2-ylmethanol |
SMILES | OCC1CO1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC120051000
|
Thermo Scientific Chemicals
120051000 |
100 mL | Glass bottle |
Each for $105.24
|
|
|||||
AC120055000
|
Thermo Scientific Chemicals
120055000 |
500 mL | Glass bottle |
Each for $379.46
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
556-52-5 | |
74.08 | |
CTKINSOISVBQLD-UHFFFAOYNA-N | |
11164 | |
oxiran-2-ylmethanol |
C3H6O2 | |
MFCD00005147,MFCD00074873 | |
glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol | |
CHEBI:30966 | |
OCC1CO1 |
Specifications
556-52-5 | |
Beige-Yellow to White | |
160.0°C to 161.0°C | |
Authentic | |
C3H6O2 | |
MFCD00005147,MFCD00074873 | |
17,104 | |
15,4525 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
OCC1CO1 | |
74.08 | |
4.0 mPa.s (20°C) | |
74.08 | |
Powder, Crystals or Chunks |
-54.0°C | |
1.1100g/mL | |
71°C | |
Glass bottle | |
1.4300 to 1.4340 | |
100 mL | |
1.11 | |
glycidol, oxiranemethanol, 2,3-epoxy-1-propanol, epihydrin alcohol, glycide, oxiranylmethanol, 3-hydroxypropylene oxide, 1-propanol, 2,3-epoxy, 2,3-epoxypropanol, glycidyl alcohol | |
CTKINSOISVBQLD-UHFFFAOYNA-N | |
oxiran-2-ylmethanol | |
11164 | |
CHEBI:30966 | |
96% | |
Glycidol |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
May cause cancer.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
GHS Signal Word: Danger
EINECSNumber : 209-128-3
RUO – Research Use Only