missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 120101000

 View more versions of this product

Catalog No. AC120101000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glycine methyl ester hydrochloride
5680-79-5
White
98%
C3H8ClNO2
MFCD00012870
04,34
Solubility in water: >1000g/L water (20 c)
[H+].[Cl-].COC(=O)CN
125.55
125.56
Crystals or Powder
98%
173°C to 177°C
Authentic
Plastic bottle
H2NCH2CO2CH3·HCl
100 g
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
COQRGFWWJBEXRC-UHFFFAOYSA-N
methyl 2-aminoacetate;hydrochloride
122755
98%

Chemical Identifiers

5680-79-5
125.55
COQRGFWWJBEXRC-UHFFFAOYSA-N
122755
[H+].[Cl-].COC(=O)CN
C3H8ClNO2
MFCD00012870
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
methyl 2-aminoacetate;hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-139-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only