missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

$100.05 - $100.05

Chemical Identifiers

CAS 5680-79-5
Molecular Formula C3H8ClNO2
Molecular Weight (g/mol) 125.55
MDL Number MFCD00012870
InChI Key COQRGFWWJBEXRC-UHFFFAOYSA-N
Synonym glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
PubChem CID 122755
IUPAC Name methyl 2-aminoacetate;hydrochloride
SMILES [H+].[Cl-].COC(=O)CN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120101000
SDS View Documents
Thermo Scientific Chemicals
120101000
100 g Plastic bottle
Each for $100.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5680-79-5
125.55
COQRGFWWJBEXRC-UHFFFAOYSA-N
122755
[H+].[Cl-].COC(=O)CN
C3H8ClNO2
MFCD00012870
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
methyl 2-aminoacetate;hydrochloride

Specifications

5680-79-5
White
98%
C3H8ClNO2
MFCD00012870
04,34
Solubility in water: >1000g/L water (20 c)
[H+].[Cl-].COC(=O)CN
125.55
125.56
Crystals or Powder
173°C to 177°C
Authentic
Plastic bottle
H2NCH2CO2CH3·HCl
100 g
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
COQRGFWWJBEXRC-UHFFFAOYSA-N
methyl 2-aminoacetate;hydrochloride
122755
98%
Glycine methyl ester hydrochloride, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-139-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only