missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

$304.97 - $5568.01

Chemical Identifiers

CAS 15283-45-1
Molecular Formula AuNa3O6S4
Molecular Weight (g/mol) 490.17
MDL Number MFCD00046176
InChI Key FJCMCHNDCIYVST-UHFFFAOYSA-J
Synonym sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4
PubChem CID 27176
IUPAC Name gold(1+) trisodium disulfanidesulfonate
SMILES [Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3974103
View Documents
Thermo Scientific Chemicals
03974103
1 g
Each for $304.97
 
AA3974106
View Documents
Thermo Scientific Chemicals
03974106
5 g
Each for $1,303.89
 
AA3974114
View Documents
Thermo Scientific Chemicals
03974114
25 g
Each for $5,568.01
 
Description

Description

Gold(I) sodium thiosulfate hydrate is useful in organogold compounds. Further, it serves as an antirheumatic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15283-45-1
490.17
FJCMCHNDCIYVST-UHFFFAOYSA-J
27176
[Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O
AuNa3O6S4
MFCD00046176
sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4
gold(1+) trisodium disulfanidesulfonate

Specifications

(decomposition)
15283-45-1
1 g
AuNa3O6S4
sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4
FJCMCHNDCIYVST-UHFFFAOYSA-J
gold(1+) trisodium disulfanidesulfonate
27176
White powder
Gold(I) sodium thiosulfate hydrate
White
Powder
99.9% (Metal basis)
MFCD00046176
Soluble in water. Insoluble in ethanol and most other organic solvents.
[Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O
490.17
490.19 (anhydrous)
3.09
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 239-324-4

RTECSNumber : CK6235000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only