missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Pulp/Aluminum Caps
For mixing, measuring and storage
Supplier: Qorpak™ GLC01459
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
1 oz., 30mL | |
1 oz. | |
1.34 in. | |
Bulk Pack | |
Black | |
2.68 in. | |
Phenolic | |
33 mm |
30 mL | |
Clear | |
Glass | |
432/Cs. | |
68 mm | |
Pulp/Aluminum Foil | |
33-400 mm | |
Screw Cap |