missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Pulp/Aluminum Caps
For mixing, measuring and storage
$145.03 - $1515.59
Specifications
Color | Clear |
---|---|
Material | Glass |
Closure Color | Black |
Closure Liner | Pulp/Aluminum Foil |
Closure Material | Phenolic |
Catalog Number | Mfr. No. | Capacity | Packaging | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Packaging | Quantity | Price | Quantity | |||
02991856
|
Qorpak™
GLC01459 |
1 oz., 30mL | Bulk Pack | 432/Cs. |
Case of 432 Each for $1,515.59
|
|
|||
02991772
|
Qorpak™
GLC01482 |
2 oz., 60 mL | Bulk Pack | 288/Cs. |
Case of 288 Each for $1,219.65
|
|
|||
02992127
|
Qorpak™
GLC06677 |
4 oz., 120 mL | Conventional Pack | 24/Cs. |
Case of 24 Each for $145.03
|
|
|||
02992226
|
Qorpak™
GLC01515 |
4 oz., 120 mL | Bulk Pack | 144/Cs. |
Case of 144 Each for $691.50
|
|
|||
02991790
|
Qorpak™
GLC01568 |
16 oz., 480 mL | Bulk Pack | 48/Cs. |
Case of 48 Each for $457.06
|
|
|||
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
Clear | |
Black | |
Phenolic |
Glass | |
Pulp/Aluminum Foil | |
Screw Cap |