Feather™ Stainless-Steel Blades for Dissecting Knife Handles Edge
Edge
Exceptionally strong and uniformly sharp
Supplier:  Feather™ NO22
 View more versions of this product
Catalog No. 08-918-5C
Edge
Explore available promotions
Description
Description
Exceptionally strong and uniformly sharp
  • Surgical quality
  • Sterile
  • Individually foil-wrapped
Specifications
Specifications
Stainless Steel
# 22
5.5 cm
Sterile
Blade
2.18 in.
For Handle # 4, # 6
Videos
SDS
Documents
Documents
Accessories