Feather™ Stainless-Steel Blades for Dissecting Knife Handles
Exceptionally strong and uniformly sharp
$48.63 - $50.24
Catalog Number | Mfr. No. | For Use With (Equipment) | Length (English) | Length (Metric) | Size | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Length (English) | Length (Metric) | Size | Price | Quantity | ||
08-918-5A
|
Feather™
NO20 |
For Handle # 4, # 6 | 1.56 in. | 4 cm | # 20 |
Pack of 100 Each for $48.63
|
|
||
08-916-5C
|
Feather™
NO12 |
For Handle # 3, # 5, # 7 | 1.37 in. | 3.5 cm | # 12 |
Pack of 100 Each for $50.24
|
|
||
08-918-5D
|
Feather™
NO23 |
For Handle # 4, # 6 | 1.75 in. | 4.5 cm | # 23 |
Pack of 100 Each for $48.63
|
|
||
08-916-5A
|
Feather™
NO10 |
For Handle # 3, # 5, # 7 | 1.56 in. | 4 cm | # 10 |
Pack of 100 Each for $48.63
|
|
||
08-916-5B
|
Feather™
NO11 |
For Handle # 3, # 5, # 7 | 1.56 in. | 4 cm | # 11 |
Pack of 100 Each for $49.58
|
|
||
08-918-5B
|
Feather™
NO21 |
For Handle # 4, # 6 | 1.94 in. | 5 cm | # 21 |
Pack of 100 Each for $48.63
|
|
||
08-918-5C
|
Feather™
NO22 |
For Handle # 4, # 6 | 2.18 in. | 5.5 cm | # 22 |
Pack of 100 Each for $48.63
|
|
||
08-916-5D
|
Feather™
NO15 |
For Handle # 3, # 5, # 7 | 1.37 in. | 3.5 cm | # 15 |
Pack of 100 Each for $48.63
|
|
||
Description
Exceptionally strong and uniformly sharp
- Surgical quality
- Sterile
- Individually foil-wrapped
Specifications
Stainless Steel | |
Blade | |
Sterile |