Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
Supplier:  Greiner Bio-One 450470
 View more versions of this product
Catalog No. 22-030-400
Explore available promotions
Description
Description
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
  • Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
  • Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
  • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
  • All tubes require MiniCollect funnel or capillary collection device (sold separately)
Specifications
Specifications
MiniCollect Capillary Blood Collection Tube
1 mL
Red
Serum
Yes
Small volume blood collection