Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
$183.72 - $215.55
Specifications
Product Type MiniCollect Capillary Blood Collection Tube
Closure Included Yes
For Use With (Application) Small volume blood collection
View More Specs
Products 4
Catalog Number Mfr. No. Additive Closure Color Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Additive Closure Color Capacity (Metric) Price Quantity  
22-030-400
View Documents
Greiner Bio-One
450470
Serum Red 1 mL
Pack of 100 Each for $183.72
Case of 10 Pack for $1,837.20
22-030-401
View Documents
Greiner Bio-One
450472
Serum Sep Gold 0.8 mL
Pack of 100 Each for $215.55
Case of 10 Pack for $2,155.50
22-030-405
View Documents
Greiner Bio-One
450477
Lithium Heparin Green 1 mL
Pack of 100 Each for $183.72
Case of 10 Pack for $1,837.20
22-030-406
View Documents
Greiner Bio-One
450479
Lithium Heparin Green 0.8 mL
Pack of 100 Each for $215.55
Case of 10 Pack for $2,155.50
Description
Description
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
  •  Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
  • Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
  • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
  • All tubes require MiniCollect funnel or capillary collection device (sold separately)
Specifications
Specifications
MiniCollect Capillary Blood Collection Tube
Yes
Small volume blood collection
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'