missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One Snap Caps
For recapping 13mm VACUETTE tubes
Supplier: Greiner Bio-One 371518
Specifications
Snap Cap | |
Yellow | |
Recapping 13 mm Vacuette™ tubes to reduce the risk of biohazardous contamination |
PET | |
Universal |