missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One Snap Caps
For recapping 13mm VACUETTE tubes
$193.97 - $244.46
Specifications
Product Type | Snap Cap |
---|---|
Material | PET |
Size | Universal |
For Use With (Equipment) | Recapping 13 mm Vacuette™ tubes to reduce the risk of biohazardous contamination |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
22040301
|
Greiner Bio-One
371518 |
Yellow |
Pack of 1000 Each for $244.46
Case of 5 Pack for $1,222.30
|
|
|||||
22040303
|
Greiner Bio-One
371522 |
Red |
Pack of 1000 Each for $244.46
Case of 5 Pack for $1,222.30
|
|
|||||
22040304
|
Greiner Bio-One
371523 |
Lavender |
Pack of 1000 Each for $244.46
Case of 5 Pack for $1,222.30
|
|
|||||
36-101-9902
|
Greiner Bio-One
371527 |
Pink |
Pack of 1000 Each for $243.31
Case of 5 Pack for $1,216.55
|
|
|||||
22-040-302
|
Greiner Bio-One
371519 |
Royal Blue |
Pack of 1000 Each for $236.03
Case of 5 Pack for $1,180.15
|
|
|||||
22-040-311
|
Greiner Bio-One
371517 |
Green |
Pack of 1000 Each for $193.97
Case of 5 Pack for $969.85
|
|
|||||
Specifications
Snap Cap | |
PET | |
Universal | |
Recapping 13 mm Vacuette™ tubes to reduce the risk of biohazardous contamination |